missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum foil, 1.0mm (0.04 in.) thick, annealed, 99% (metals basis), Si and Fe (combined) typically 1%

Manufacturer:  Alfa Aesar™ 14192VV

 View more versions of this product

Catalog No. AA14192VV



Specifications

Specifications

Aluminum foil
45 x 240cm
7429-90-5
(metals basis)
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum
26.982
CHEBI:33629
99%
≈729g/45x60cm
1.0mm (0.04 in.) thick, Annealed, Temper: soft
Si and Fe (combined) typically 1%
Al
aluminium, metal, powder, aluminio, metana, adom, alumina fibre, aluminium flake, dust, noral aluminium
[Al]
Odorless
5359268
Foil

Chemical Identifiers

7429-90-5
26.982
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
aluminum
Al
MFCD00134029
aluminium, metal, powder, aluminio, metana, adom, alumina fibre, aluminium flake, dust, noral aluminium
CHEBI:33629
[Al]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents