missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum foil, 0.2mm (0.008 in.) thick, Puratronic™, 99.997% (metals basis)

Manufacturer:  Alfa Aesar™ 43776GH

 View more versions of this product

Catalog No. AA43776GH



Specifications

Specifications

Aluminum foil
100 x 100mm
7429-90-5
Al
aluminium, metal, powder, aluminio, metana, adom, alumina fibre, aluminium flake, dust, noral aluminium
[Al]
Odorless
5359268
Foil
Puratronic™
≈5.4g/100x100mm
0.2mm (0.008 in.) thick, Temper: as rolled
(metals basis)
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum
26.982
CHEBI:33629
99.997%

Chemical Identifiers

7429-90-5
26.982
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
aluminum
Al
MFCD00134029
aluminium, metal, powder, aluminio, metana, adom, alumina fibre, aluminium flake, dust, noral aluminium
CHEBI:33629
[Al]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents