missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum Foil, 0.25mm (0.01 in.) thick, annealed, Puratronic™, 99.9999% (Metals basis), Alfa Aesar™

Form: Approx. 1.7g/50 x 50mm; Temper: soft

Manufacturer:  Alfa Aesar 44331FI

 View more versions of this product

Catalog No. AA44331FI



Specifications

Specifications

Aluminum
Annealed
Silver-colored foil
Al
aluminium, metal, powder, aluminio, metana, adom, alumina fibre, aluminium flake, dust, noral aluminium
Odorless
aluminum
5359268
Foil, 0.25mm (0.01 in.) thick
50 x 50mm
7429-90-5
99.9999% (Metals basis)
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
[Al]
26.982
CHEBI:33629
Silver

Chemical Identifiers

7429-90-5
26.982
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
aluminum
Al
MFCD00134029
aluminium, metal, powder, aluminio, metana, adom, alumina fibre, aluminium flake, dust, noral aluminium
CHEBI:33629
[Al]
SDS Documents