missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ Aluminum foil, 0.25mm (0.01 in.) thick, annealed, 99% (metals basis), Si and Fe (combined) typically 1%
Manufacturer: Alfa Aesar™ 14196RW
Specifications
Aluminum foil | |
30 x 50cm | |
7429-90-5 | |
(metals basis) | |
MFCD00134029 | |
XAGFODPZIPBFFR-UHFFFAOYSA-N | |
aluminum | |
26.982 | |
CHEBI:33629 | |
99% |
≈102.8g/30x50cm | |
0.25mm (0.01 in.) thick, Annealed, Temper: soft | |
Si and Fe (combined) typically 1% | |
Al | |
aluminium, metal, powder, aluminio, metana, adom, alumina fibre, aluminium flake, dust, noral aluminium | |
[Al] | |
Odorless | |
5359268 | |
Foil |
Chemical Identifiers
7429-90-5 | |
26.982 | |
XAGFODPZIPBFFR-UHFFFAOYSA-N | |
5359268 | |
aluminum |
Al | |
MFCD00134029 | |
aluminium, metal, powder, aluminio, metana, adom, alumina fibre, aluminium flake, dust, noral aluminium | |
CHEBI:33629 | |
[Al] |
Safety and Handling
TSCA : TSCA