missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum foil, 0.025mm (0.001 in.) thick, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 44233GA

 View more versions of this product

Catalog No. AA44233GA



Specifications

Specifications

Aluminum foil
50 x 200mm
7429-90-5
Al
adom
[Al]
Odorless
5359268
Foil
≈0.675g/50x200mm
0.025mm (0.001 in.) thick
(metals basis)
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum
26.982
CHEBI:33629
99.999%

Chemical Identifiers

7429-90-5
26.982
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
aluminum
Al
MFCD00134029
adom
CHEBI:33629
[Al]
Safety and Handling

Safety and Handling

EINECSNumber : 231-072-3

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents