missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum fluoride hydrate, Puratronic™, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 1062322

 View more versions of this product

Catalog No. AA1062322



Specifications

Specifications

Aluminum fluoride hydrate
(metals basis)
MFCD00003426
aluminum fluoride hydrate, aluminium fluoride hydrate, alf3.h2o, trifluoroalumane-water 1/1, aluminum fluoride hydrate, puratronic
O.F[Al](F)F
101.992
83.98 (Anhydrous)
99.99%
−22 Mesh
32287-65-3
AlF3H2O
UN3260
XTXFDUABTPNTFB-UHFFFAOYSA-K
trifluoroalumane;hydrate
16211462
Powder
Puratronic™
100g

Chemical Identifiers

32287-65-3
101.992
XTXFDUABTPNTFB-UHFFFAOYSA-K
16211462
O.F[Al](F)F
AlF3H2O
MFCD00003426
aluminum fluoride hydrate, aluminium fluoride hydrate, alf3.h2o, trifluoroalumane-water 1/1, aluminum fluoride hydrate, puratronic
trifluoroalumane;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H300-H314-H318
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

Documents