missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum fluoride, anhydrous, 99+%

Manufacturer:  Alfa Aesar™ 1149703

 View more versions of this product

Catalog No. AA1149703



Description

Description

  • In ceramics, as flux in metallurgy, in aluminum manufacture, as inhibitor of fermentation, as catalyst in organic reactions
Specifications

Specifications

Aluminum fluoride
7784-18-1
14,339
83.98
≥99%
Hygroscopic
1g
Anhydrous
MFCD00003426
Practically insoluble in water; Sparingly soluble in acids and alkalies
Powder
2.882g/mL
1291°C (sublimation)

Chemical Identifiers

7784-18-1
MFCD00003426
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : BD0725000

TSCA : TSCA

Documents