missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum fluoride, anhydrous, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 4443518

 View more versions of this product

Catalog No. AA4443518



Specifications

Specifications

Aluminum fluoride
7784-18-1
AlF3
14,339
83.98
99.99%
Hygroscopic
50g
Anhydrous
(metals basis)
MFCD00003426
Slightly soluble in water.
Powder
2.882 g/mL
1291°C (sublimation)

Chemical Identifiers

7784-18-1
AlF3
MFCD00003426
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

EINECSNumber : 232-051-1

RTECSNumber : BD0725000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents