missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum Flake, 11 micron, 99.7% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 45031A1

 View more versions of this product

Catalog No. AA45031A1



Specifications

Specifications

Aluminum
7429-90-5
2.7g/cm3
99.7% (Metals basis)
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
[Al]
26.982
CHEBI:33629
Moisture sensitive
1kg
Silver Gray flakes
400°C (752°F)
Al
aluminium, metal, powder, aluminio, metana, adom, alumina fibre, aluminium flake, dust, noral aluminium
Odorless
aluminum
5359268
Flakes
Silver to Gray

Chemical Identifiers

7429-90-5
26.982
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
aluminum
Al
MFCD00134029
aluminium, metal, powder, aluminio, metana, adom, alumina fibre, aluminium flake, dust, noral aluminium
CHEBI:33629
[Al]
Safety and Handling

Safety and Handling

Keep container tightly closed and dry.

Contact with water liberates highly flammable gases. Spontaneously flammable in air.

DOTInformation : DOT Class: 4.3, Packaging Group: II

EINECSNumber : 231-072-3

SDS Documents