missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Aluminum etchant, Thermo Scientific Chemicals
Standard Aluminum Etchant For Use On Silicon Devices and Other Microelectronic Applications
$150.63 - $150.63
Specifications
Chemical Name or Material | Aluminum etchant |
---|---|
MDL Number | MFCD08064399 |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA44581R0
|
Thermo Scientific Chemicals
044581R0 |
1 qt. |
N/A
|
|
|||||
Description
Standard aluminum etchant for use on silicon devices and other microelectronic applications
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Aluminum etchant | |
MFCD08064399 | |
Liquid |
RUO – Research Use Only