missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum dihydrogen phosphate, 50% w/w aqueous solution, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 42858A7

 View more versions of this product

Catalog No. AA42858A7



Description

Description

Aluminum dihydrogen phosphate is used as intermediates and solid separation agents. It is also used in petroleum, chemical industry, shipbuilding, space technology, electric industry, high-temperature kilns, heat treatment resistance furnaces and electric insulation. Further, it is used as inorganic coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Aluminum dihydrogen phosphate
13530-50-2
5 kg
AlH6O12P3
aluminum dihydrogen phosphate, aluminum phosphate monobasic, prim-aluminum phosphate, aluminum tris dihydrogen phosphate, phosphoric acid, aluminum salt 3:1, aluminum hydrogen phosphate, aluminumdihydrogen phosphate, aluminum phosphate,monobasic, aluminum trisdihydrogenphosphate, aluminum dihydrogen orthophosphate
RGPUVZXXZFNFBF-UHFFFAOYSA-K
aluminum;dihydrogen phosphate
9818426
Colorless liquid
Colorless
Liquid
50% w/w aqueous solution
MFCD00014152
Soluble in water and ethanol. Insoluble in acetone.
[Al+3].OP(O)([O-])=O.OP(O)([O-])=O.OP(O)([O-])=O
317.94
317.94
1.5 g/cm3

Chemical Identifiers

13530-50-2
317.94
RGPUVZXXZFNFBF-UHFFFAOYSA-K
9818426
[Al+3].OP(O)([O-])=O.OP(O)([O-])=O.OP(O)([O-])=O
AlH6O12P3
MFCD00014152
aluminum dihydrogen phosphate, aluminum phosphate monobasic, prim-aluminum phosphate, aluminum tris dihydrogen phosphate, phosphoric acid, aluminum salt 3:1, aluminum hydrogen phosphate, aluminumdihydrogen phosphate, aluminum phosphate,monobasic, aluminum trisdihydrogenphosphate, aluminum dihydrogen orthophosphate
aluminum;dihydrogen phosphate
Safety and Handling

Safety and Handling

EINECSNumber : 215-691-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only