Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum di(sec-butoxide)acetoacetic ester chelate, tech., Al 8.4% min

$64.03 - $64.03

Chemical Identifiers

Chemical Name or Material Aluminum di (sec-butoxide)acetoacetic ester chelate
CAS 24772-51-8
Purity Grade Technical Grade
Molecular Formula C14H27AlO5
Formula Weight 302.3
MDL Number MFCD00084762
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA8934936

 
alfa aesar™
8934936
500g Each for $64.03
Description & Specifications

Specifications

Formula Weight 302.3
Chemical Name or Material Aluminum di (sec-butoxide)acetoacetic ester chelate
Concentration Al 8.4% min
Sensitivity Moisture Sensitive
TSCA TSCA
Purity Grade Technical Grade
Physical Form Liquid
Molecular Formula C14H27AlO5
MDL Number MFCD00084762
CAS 24772-51-8
Quantity 500g
Flash Point 76°C (169°F)
Health Hazard 1 Warning
Density 1.03g/mL
Health Hazard 2 GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Alcohol-like
Safety and Handling

Warning

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.