missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum di(isopropoxide)acetoacetic ester chelate, Al 9.6% min, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 08935022

 View more versions of this product

Catalog No. AA8935022



Description

Description

Aluminum di(isopropoxide)acetoacetic ester chelate is used as a gel ink. It finds application in paint to give better drying, luster and hardness, and improve the performance as surface wrinkling. It is also used as a crosslinking agent in epoxy, alkyd and polyester resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Aluminum di (isopropoxide)acetoacetic ester chelate
1.05 g/mL
Alcohol-like
MFCD00135521
UN1993
274.3
Liquid
14782-75-3
50°C (122°F)
C12H23AlO5
100 g
Moisture sensitive
≥9.6%

Chemical Identifiers

14782-75-3
C12H23AlO5
MFCD00135521
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 238-851-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only