missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum di(isopropoxide)acetoacetic ester chelate, Al 9.6% min

Manufacturer:  Alfa Aesar™ 8935036

 View more versions of this product

Catalog No. AA8935036



Specifications

Specifications

Aluminum di (isopropoxide)acetoacetic ester chelate
500g
50°C (122°F)
Alcohol-like
C12H23AlO5
UN1993
Liquid
Al 9.6% min
14782-75-3
1.05g/mL
Moisture Sensitive
MFCD00135521
274.3
≥9.6%

Chemical Identifiers

14782-75-3
C12H23AlO5
MFCD00135521
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
F IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA

SDS Documents