Thermo Scientific™ Aluminum Crimp Seals with Prefitted Septa for 12 x 32mm Vials
You can count on a secure, leak-resistant seal with Thermo Scientific™ National Aluminum Crimp Seals.
$50.68 - $109.91
Specifications
Products 3
Catalog Number Mfr. No. Type Closure Color Septum Price Quantity  
Catalog Number Mfr. No. Type Closure Color Septum Price Quantity  
06-406-19T
View Documents
Thermo Scientific™
C40113A
Crimp Cap with Septum Silver Polypropylene septa Pack of 100 Each for $50.68
Case of 10 Pack for $506.80
03-376-411
View Documents
Thermo Scientific™
C40114Y
Crimp Cap Seal Yellow Prefitted Red PTFE/White Silicone Septa Pack of 100 Each for $63.54
03-378-343
View Documents
Thermo Scientific™
C40115A
Crimp Cap with Septum Silver Black Viton Elastomer Pack of 100 Each for $109.91
Description
Description
You can count on a secure, leak-resistant seal with Thermo Scientific™ National Aluminum Crimp Seals.
  • Anodized aluminum seals fit all 11mm crimp-top vials
  • Seal 2000™ material is specially formulated for improved background performance over PTFE/red rubber
  • Viton™ septa perform well with ECD detectors and are resistant to many aggressive solvents
Specifications
Specifications
0.0
icon-pdf.png