missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Aluminum Crimp Seals with Prefitted Septa for 12 x 32mm Vials

You can count on a secure, leak-resistant seal with Thermo Scientific™ National Aluminum Crimp Seals.

Manufacturer:  Thermo Scientific™ C40113A

 View more versions of this product

Catalog No. 06-406-19T




Description

Description

You can count on a secure, leak-resistant seal with Thermo Scientific™ National Aluminum Crimp Seals.

  • Anodized aluminum seals fit all 11mm crimp-top vials
  • Seal 2000™ material is specially formulated for improved background performance over PTFE/red rubber
  • Viton™ septa perform well with ECD detectors and are resistant to many aggressive solvents
Specifications

Specifications

Silver
Silver
Polypropylene septa
Aluminum
11 mm
Polypropylene septa; Silver Aluminum Seal
Polypropylene
National™
Crimp
12x32 Vials
SDS
Documents
Product Certifications