Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum cobalt isopropoxide, 99.9% (metals basis), 10% w/v in isopropanol

Chemical Identifiers

Chemical Name or Material Aluminum cobalt isopropoxide
CAS 70504-56-2
Formula Weight 585.15
MDL Number MFCD00143589
UN Number UN1219
Synonym Cobalt aluminum isopropoxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA36593AC

  SDS
alfa aesar™
36593AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA36593AE

  SDS
alfa aesar™
36593AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 585.15
Chemical Name or Material Aluminum cobalt isopropoxide
Concentration 10% w/v in isopropanol
Sensitivity Moisture Sensitive
TSCA TSCA
Boiling Point 140°C (0.6mmHg)
Physical Form Liquid Suspension
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00143589
CAS 70504-56-2
Quantity 25mL
UN Number UN1219
Synonym Cobalt aluminum isopropoxide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H341-H315-H319-H336
Highly flammable liquid and vapor.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H341-H315-H319-H336
Highly flammable liquid and vapor.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.