missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum chloride, ultra dry, 99.99% (metals basis), Alfa Aesar

Manufacturer:  Alfa Aesar 1455206

 View more versions of this product

Catalog No. AA1455206



Specifications

Specifications

Aluminum chloride
7446-70-0
AlCl3
Yellow to gray powder
133.34
2.44g/cm3
Gray to Yellow
ultra dry
99.99% (Metal basis)
MFCD00003422
aluminium chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminium
Powder
Pungent
5g

Chemical Identifiers

7446-70-0
AlCl3
MFCD00003422
aluminium chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminium
Safety and Handling

Safety and Handling

H314

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 231-208-1

Documents