Learn More
Aluminum chloride hexahydrate, Reagent Grade, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 012297A3
Description
Pharmaceutical preparations of aluminum chloride hexahydrate are used to treat hyperhidrosis, a condition involving excessive sweating. It is also employed in deodorants, antiperspirant preparations, dyeing fabrics, refining crude oil, textile finishing, manufacturing parchment paper, preserving the wood and disinfecting stables. It also finds use as a reagent in organic synthesis. Aluminum chloride hexahydrate is also useful in water treatment process, medicine and cosmetic products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Aluminum chloride hexahydrate | |
White | |
Crystals | |
AlCl3H12O6 | |
Highly soluble in water. Soluble in alcohol,ether,glycerol,propylene glycol and acetonitrile. | |
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-] | |
241.42 | |
Reagent | |
2.398 |
100°C (decomposition) | |
7784-13-6 | |
2 kg | |
MFCD00149134 | |
JGDITNMASUZKPW-UHFFFAOYSA-K | |
aluminium(3+) hexahydrate trichloride | |
241.43 (133.34 Anhydrous) | |
Odorless |
Chemical Identifiers
7784-13-6 | |
241.42 | |
JGDITNMASUZKPW-UHFFFAOYSA-K |
AlCl3H12O6 | |
MFCD00149134 | |
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-] |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : DOT Class: 8, Packaging Group: III
EINECSNumber : 231-208-1
RTECSNumber : BD0530000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only