Please login to your online account to display your discounted pricing

Aluminum Chloride, Hexahydrate, Reagent Grade, LabChem™

$118.86 - $118.86

Chemical Identifiers

Chemical Name or Material Aluminum Chloride
CAS 7784-13-6
CAS Max % 100
Name Note Hexahydrate
Purity Grade Reagent Grade
Linear Formula AlCl3·6H2O
Formula Weight 241.43
UN Number UN1726
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC108101

 
lab chem
LC108101
500g Each for $118.86
Description & Specifications

Specifications

Formula Weight 241.43
Melting Point 100°C
Boiling Point 182°C
Chemical Name or Material Aluminum Chloride
CAS Max % 100
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
Avoid breathing dust, fume.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical attention.
Take off contaminated clothing and wash it before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical attention.
Store in a well-ventilated place.
Keep container tightly closed.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Purity Grade Reagent Grade
Density 2.398g/cm3
Specific Gravity 2.398
Storage Information Room Temperature
Color White
Physical Form Crystals
UN Number UN1726
Health Hazard 2 GHS H Statement
Causes skin irritation and serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life.
Quantity 500g
Linear Formula AlCl3·6H2O
Decomposition Information Fume; Hydrogen chloride
Solubility Soluble in water
CAS 7784-13-6
Name Note Hexahydrate
Packaging Poly Bottle
Identification Passes Test
Safety and Handling

Warning

GHS P Statement
Avoid breathing dust, fume.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical attention.
Take off contaminated clothing and wash it before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical attention.
Store in a well-ventilated place.
Keep container tightly closed.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation and serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life.