missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum chloride hexahydrate, Puratronic™, 99.9995% (metals basis)

Manufacturer:  Alfa Aesar™ 1062206

 View more versions of this product

Catalog No. AA1062206



Specifications

Specifications

Aluminum chloride hexahydrate
(metals basis)
UN3260
241.43 (133.34 Anhydrous)
99.9995%
2.398g/mL
White
2.5
7784-13-6
MFCD00149134
14,337
Crystalline Aggregates
Puratronic™
Odorless
100°C (decomposition)
5g

Chemical Identifiers

7784-13-6
MFCD00149134
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

RTECSNumber : BD0530000

TSCA : TSCA

SDS Documents