Please login to your online account to display your discounted pricing

Aluminum Chloride Hexahydrate (Crystalline/Certified), Fisher Chemical

$313.62 - $313.62

Chemical Identifiers

Chemical Name or Material Aluminum Chloride Hexahydrate
CAS 7784-13-6
Purity Grade Certified Grade
Physical Form Solid
Molecular Formula AlCl3 · 6 H2O
Formula Weight 241.43
MDL Number MFCD00149134
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

A573-500

  Accessories SDS
fisher scientific company
FLA573500
Glass Bottle 500g Each for $313.62
Case of 6 Each for $1,881.72
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 241.43
Melting Point 181°C
Quantity 500g
DOT Information DOT Class 8, : Corrosive
Physical Form Solid
Viscosity [NULL]
Color White
pH 2.0
Molecular Formula AlCl3 · 6 H2O
Density 2.39g/cm³
Identification Pass Test
Insoluble Matter 0.005% max.
Alkalis and Alkaline Earths 0.20% max.
Iron (Fe) 5ppm max.
Ammonia 0.010% max.
Chemical Name or Material Aluminum Chloride Hexahydrate
Free Acid 0.10% max.
Sulfate 0.010% max.
MDL Number MFCD00149134
ChemAlert Storage Symbol White
Heavy Metals (as Pb) 5ppm max.
Packaging Glass Bottle
CAS 7784-13-6

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.