missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum chloride, anhydrous, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 4447022

 View more versions of this product

Catalog No. AA4447022



Specifications

Specifications

Aluminum chloride
7446-70-0
AlCl3
UN1726
aluminium chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminium
99.999%
−10 Mesh
Moisture Sensitive
2.4
Anhydrous
(metals basis)
MFCD00003422
14,337
133.34
2.44g/mL
Pungent
190°C (sublimation)
100g

Chemical Identifiers

7446-70-0
AlCl3
MFCD00003422
aluminium chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminium
Safety and Handling

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ALUMINUM CHLORIDE, ANHYDROUS

RTECSNumber : BD0525000

TSCA : TSCA

SDS Documents