missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum chloride, anhydrous, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 4447014

 View more versions of this product

Catalog No. AA4447014



Specifications

Specifications

Aluminum chloride
7446-70-0
AlCl3
UN1726
alumimum chloride
133.34
99.999%
−10 Mesh
Moisture sensitive
2.4
Anhydrous
(metals basis)
MFCD00003422
14,337
Soluble in benzene,carbon tetrachloride,chloroform,hydrochloric acid,ether,benzophenone,nitrobenzene and ethanol.
-10 Mesh
2.44 g/mL
Pungent
190°C (sublimation)
25g

Chemical Identifiers

7446-70-0
AlCl3
MFCD00003422
alumimum chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ALUMINUM CHLORIDE, ANHYDROUS

EINECSNumber : 231-208-1

RTECSNumber : BD0525000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents