missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum chloride, anhydrous, 99.985% (metals basis), Alfa Aesar™

$52.53 - $214.32

Chemical Identifiers

CAS 7446-70-0
Molecular Formula AlCl3
Molecular Weight (g/mol) 133.33
MDL Number MFCD00003422
InChI Key VSCWAEJMTAWNJL-UHFFFAOYSA-K
Synonym alumimum chloride, aluminium chloride anhydrous, aluminiumchloride, aluminum chlorid, aluminum cloride, aluminum iii chloride, aluminum-chloride, aluminumchloride, chloride aluminium, trichloroaluminium
IUPAC Name aluminium(3+) trichloride
SMILES [Al+3].[Cl-].[Cl-].[Cl-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA8848806
View Documents
Thermo Fisher Scientific
8848806
5g
Each for $52.53
 
AA8848818
View Documents
Thermo Fisher Scientific
8848818
50g
Each for $82.55
 
AA8848830
View Documents
Thermo Fisher Scientific
8848830
250g
Each for $214.32
 
Description

Description

Specifications

Chemical Identifiers

7446-70-0
133.33
VSCWAEJMTAWNJL-UHFFFAOYSA-K
aluminium(3+) trichloride
AlCl3
MFCD00003422
alumimum chloride, aluminium chloride anhydrous, aluminiumchloride, aluminum chlorid, aluminum cloride, aluminum iii chloride, aluminum-chloride, aluminumchloride, chloride aluminium, trichloroaluminium
[Al+3].[Cl-].[Cl-].[Cl-]

Specifications

7446-70-0
AlCl3
Yellow to gray
Soluble in water,hydrochloric acid and ethanol. Slightly soluble in benzene.
[Al+3].[Cl-].[Cl-].[Cl-]
133.33
Powder
Pungent
Gray to Yellow
5g
99.985% (Metal basis)
MFCD00003422
alumimum chloride, aluminium chloride anhydrous, aluminiumchloride, aluminum chlorid, aluminum cloride, aluminum iii chloride, aluminum-chloride, aluminumchloride, chloride aluminium, trichloroaluminium
VSCWAEJMTAWNJL-UHFFFAOYSA-K
aluminium(3+) trichloride
133.34
2.44 g/cm3
Moisture sensitive
190° (sublimation)
Aluminum chloride
Safety and Handling

Safety and Handling

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 231-208-1

RTECSNumber : BD0525000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS