missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum chloride, anhydrous, 99.985% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 8848830

 View more versions of this product

Catalog No. AA8848830



Specifications

Specifications

Aluminum chloride
99.985% (Metal basis)
MFCD00003422
aluminium chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminium
Powder
Gray to Yellow
7446-70-0
AlCl3
Yellow to gray
133.34
Pungent
250g

Chemical Identifiers

7446-70-0
AlCl3
MFCD00003422
aluminium chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminium
Safety and Handling

Safety and Handling

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 231-208-1

SDS Documents