missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum chloride, anhydrous, >95%, Alfa Aesar™

Manufacturer:  Alfa Aesar 89914D9

 View more versions of this product

Catalog No. AA89914D9



Specifications

Specifications

Aluminum chloride
>95%
MFCD00003422
aluminium chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminium
Powder
Gray to Yellow
7446-70-0
AlCl3
Yellow to gray
133.34
Pungent
5 x 1kg

Chemical Identifiers

7446-70-0
AlCl3
MFCD00003422
aluminium chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminium
Safety and Handling

Safety and Handling

H314

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 231-208-1

SDS Documents