Please login to your online account to display your discounted pricing

Aluminum chloride, 6.0N Standardized Solution, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Aluminum chloride
Molecular Formula AlCl3
Formula Weight 133.34
MDL Number MFCD00003422
UN Number UN2581
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA35568AK

  SDS
alfa aesar
35568AK
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 133.34
Quantity 250mL
Chemical Name or Material Aluminum chloride
TSCA TSCA
Odor Odorless
Physical Form Liquid
Molecular Formula AlCl3
MDL Number MFCD00003422
UN Number UN2581
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Concentration 6.0N Standardized Solution
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALUMINUM CHLORIDE, SOLUTION
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.