missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Aluminum chloride, 0.5M aq. soln., Thermo Scientific™
$122.30 - $122.30
Chemical Identifiers
CAS | 7784-13-6 |
---|---|
Molecular Formula | AlCl3H12O6 |
Molecular Weight (g/mol) | 241.42 |
MDL Number | MFCD00149134 |
InChI Key | JGDITNMASUZKPW-UHFFFAOYSA-K |
SMILES | O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ61582AE
|
Thermo Scientific Chemicals
J61582AE |
100 mL |
N/A
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7784-13-6 | |
241.42 | |
JGDITNMASUZKPW-UHFFFAOYSA-K |
AlCl3H12O6 | |
MFCD00149134 | |
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-] |
Specifications
Colorless | |
Liquid | |
AlCl3H12O6 | |
UN2581 | |
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-] | |
241.42 | |
0.5M aq. soln. |
7784-13-6 | |
100 mL | |
MFCD00149134 | |
JGDITNMASUZKPW-UHFFFAOYSA-K | |
aluminium(3+) hexahydrate trichloride | |
133.34 | |
Aluminum chloride |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALUMINUM CHLORIDE, SOLUTION
TSCA : Yes
Recommended Storage : Ambient temperatures