Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum carbonate, basic

$37.05 - $114.66

Chemical Identifiers

Chemical Name or Material Aluminum carbonate
CAS 1339-92-0
Name Note basic
MDL Number MFCD00065407
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1299418

  SDS
alfa aesar™
A1299418
50g Each for $37.05

AAA1299430

  SDS
alfa aesar™
A1299430
250g Each for $114.66
Description & Specifications

Specifications

MDL Number MFCD00065407
CAS 1339-92-0
Chemical Name or Material Aluminum carbonate
TSCA TSCA
Name Note basic
Quantity 50g

Applications
Aluminum carbonate is a popular antacid that is also a phosphate binder. It usually forms clumps that bind phosphorous in aqueous suspensions and reduces acidity and calculi.

Solubility
Insoluble in water.

Notes
Keep containers tightly closed in a dry, cool and well-ventilated place. Store at room temperature. Incompatible with strong acids and strong oxidizing agents.