missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum carbide, 99+% (metals basis)

Manufacturer:  Alfa Aesar™ 1403830

 View more versions of this product

Catalog No. AA1403830



Specifications

Specifications

Aluminum carbide
(metals basis)
MFCD00003425
14,335
CAVCGVPGBKGDTG-UHFFFAOYSA-N
alumanylidynemethyl(alumanylidynemethylalumanylidenemethylidene)alumane
16685054
≥99%
−325 Mesh
Moisture Sensitive
2100°C (>2200°C (decomposition))
1299-86-1
C3Al4
UN1394
aluminum carbide, 1,3,5,7-tetraaluminahepta-3,4-dien-1,6-diyne, aluminum carbide,al4c3, al4c3 aluminium carbide, aluminum carbide, al4c3, aluminum carbide, powder, ~325 mesh
C(#[Al])[Al]=C=[Al]C#[Al]
143.959
143.96
2.36g/mL
typically ≤10 Micron
Gray
250g

Chemical Identifiers

1299-86-1
143.959
CAVCGVPGBKGDTG-UHFFFAOYSA-N
16685054
C(#[Al])[Al]=C=[Al]C#[Al]
C3Al4
MFCD00003425
aluminum carbide, 1,3,5,7-tetraaluminahepta-3,4-dien-1,6-diyne, aluminum carbide,al4c3, al4c3 aluminium carbide, aluminum carbide, al4c3, aluminum carbide, powder, ~325 mesh
alumanylidynemethyl(alumanylidynemethylalumanylidenemethylidene)alumane
Safety and Handling

Safety and Handling

GHS H Statement
H228-H261-H315-H319-H335
Flammable solid.
In contact with water releases flammable gas.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P231+P232-P261-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ALUMINUM CARBIDE

TSCA : TSCA

Documents