missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum bromide, ultra dry, 99.999% (metals basis), Alfa Aesar™

Used as a catalyst.

Manufacturer:  Alfa Aesar 4716814

 View more versions of this product

Catalog No. AA4716814



Specifications

Specifications

Aluminum bromide
265°C
25g
AlBr3
UN1725
Powder
Plastic bottle; Ampouled under argon
97.5°C
7727-15-3
MFCD00003421
266.71
99.999%

Chemical Identifiers

7727-15-3
AlBr3
MFCD00003421
Safety and Handling

Safety and Handling

TSCA : Yes

SDS Documents