missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminium bromide, 98+%, extra pure, anhydrous, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 198415000

Catalog No. AC198415000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Lewis acid
Specifications

Specifications

Aluminium bromide
500g
AlBr3
01,22; 02,19; 03,7; 04,10; 05,10; 06,17; 09,11
Solubility in water: reacts. Other solubilities: soluble in alcohol,carbon disulfide,benzene,,toluene,ether,nitrobenzene and xylene
266.7
Extra Pure
7727-15-3
98% min. (Complexometry)
AlBr3
15,328
266.7
98+%
Glass bottle
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 231-779-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only