Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum arsenide, 99.5% (metals basis)

In rectifiers, transistors, thermistors

Chemical Identifiers

Chemical Name or Material Aluminum arsenide
CAS 22831-42-1
Formula Weight 101.9
MDL Number MFCD00046166
UN Number UN1557
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1452206

  SDS
alfa aesar™
1452206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1452214

  SDS
alfa aesar™
1452214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 101.9
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
Health Hazard 3 GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Melting Point 1740°C
Mesh Size −20 Mesh
Assay Percent Notes (metals basis)
MDL Number MFCD00046166
CAS 22831-42-1
TSCA TSCA
Chemical Name or Material Aluminum arsenide
Physical Form Powder
Health Hazard 1 Danger
Density 3.81g/mL
UN Number UN1557
Health Hazard 2 GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Assay 99.5%
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.