missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum AA Standard, 1mL = 1mg Al (1,000ppm Al) in 3% HCl, Ricca Chemical

Manufacturer:  Ricca Chemical Company RIAAL1KH500

 View more versions of this product

Catalog No. AAL1KH-500



Specifications

Specifications

Aluminum AA Standard
93.0,4.41,0.09
AlH18N3O18
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
375.129
Liquid
1,000ppm Al
Colorless
7784-27-2,7647-01-0,7429-90-5,7732-18-5
97.0,4.59,0.1
SWCIQHXIXUMHKA-UHFFFAOYSA-N
aluminum;trinitrate;nonahydrate
24567
Poly natural
1 mL = 1 mg Al
500mL

Chemical Identifiers

7784-27-2
375.129
24567
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
AlH18N3O18
SWCIQHXIXUMHKA-UHFFFAOYSA-N
aluminum;trinitrate;nonahydrate
SDS Documents