missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum 2,4-pentanedionate, 99.995+% (metals basis)

Manufacturer:  Alfa Aesar™ 4453814

 View more versions of this product

Catalog No. AA4453814



Specifications

Specifications

Aluminum 2,4-pentanedionate
314°C to 316°C
25g
(metals basis)
MFCD00000013
4157942
324.31
≥99.995%
1.27 g/mL
192°C to 194°C
13963-57-0
C15H21AlO6
UN3467
Aluminum acetylacetonate
Powder

Chemical Identifiers

13963-57-0
C15H21AlO6
MFCD00000013
Aluminum acetylacetonate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H300-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 237-741-6

RTECSNumber : BD2230000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents