Please login to your online account to display your discounted pricing

Aluminon, ACS reagent, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC401500250

 View more versions of this product

Catalog No. AC401500250

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Aluminon
CAS 569-58-4
Health Hazard 1 Warning
Packaging Glass bottle
Purity Grade ACS Reagent Grade
Molecular Formula C22H23N3O9
Formula Weight 473.44
Synonym Aurintricarboxylic acid, triammonium salt
Quantity 25g

Specifications

Chemical Name or Material Aluminon
CAS 569-58-4
Purity Grade ACS Reagent Grade
Molecular Formula C22H23N3O9
Formula Weight 473.44
Synonym Aurintricarboxylic acid, triammonium salt
Accessories

Warning

SDS & Safety Documents