Please login to your online account to display your discounted pricing

Aluminon, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC103065000

 View more versions of this product

Catalog No. AC103065000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 473.44
Physical Form Crystalline Powder
Quantity 500g
Color Deep red
Solubility Solubility in water: 800g/L (20°C). Other solubilities: slightly soluble in ethanol, insoluble in petroleum ether
Merck Index 15, 318
Beilstein 10, IV, 4161
Packaging Glass bottle
Infrared Spectrum Authentic
Suitability suitable for determination of aluminium
Chemical Name or Material Aluminon
Molecular Formula C22H23N3O9
CAS 569-58-4
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
MDL Number MFCD00040925
Sulfated Ash 0.2% max.
Synonym Aurintricarboxylic acid, triammonium salt

Specifications

Chemical Name or Material Aluminon
CAS 569-58-4
Molecular Formula C22H23N3O9
Formula Weight 473.44
MDL Number MFCD00040925
Synonym Aurintricarboxylic acid, triammonium salt
Accessories

Warning

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents