Please login to your online account to display your discounted pricing

Aluminium Tri-sec-butoxide, Acros Organics


Quantity 800mL

Manufacturer: thermo fisher scientific  AC381598000

 View more versions of this product

Catalog No. AC381598000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 246.33
Quantity 800mL
Density 1.237g/mL
Linear Formula (C2H5CH(CH3)O)3Al
CAS 75-09-2
Chemical Name or Material Aluminium tri-sec-butoxide
MDL Number MFCD00009327
CAS 2269-22-9
Molecular Formula C12H27AlO3

Specifications

Chemical Name or Material Aluminium tri-sec-butoxide
CAS 2269-22-9
CAS 75-09-2
Molecular Formula C12H27AlO3
Linear Formula (C2H5CH(CH3)O)3Al
Formula Weight 246.33
MDL Number MFCD00009327