Please login to your online account to display your discounted pricing

Aluminium Tri-sec-butoxide, Acros Organics

Chemical Identifiers

Chemical Name or Material Aluminium tri-sec-butoxide
CAS 2269-22-9
CAS 75-09-2
Molecular Formula C12H27AlO3
Linear Formula (C2H5CH(CH3)O)3Al
Formula Weight 246.33
MDL Number MFCD00009327
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC381598000

  SDS
thermo fisher scientific
AC381598000
800mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC381591000

  SDS
thermo fisher scientific
AC381591000
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 246.33
Quantity 800mL
Density 1.237g/mL
Chemical Name or Material Aluminium tri-sec-butoxide
Molecular Formula C12H27AlO3
Linear Formula (C2H5CH(CH3)O)3Al
MDL Number MFCD00009327
CAS 2269-22-9
CAS 75-09-2

Lewis acid