Please login to your online account to display your discounted pricing

Aluminium oxide, weakly acidic, Brockmann I, For chromatography, ACROS Organics™


Quantity 5kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC392520050

 View more versions of this product

Catalog No. AC392520050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 101.96
Physical Form Powder
Quantity 5kg
Color White to off-white
Merck Index 15, 350
Purity Grade For chromatography
pH 5.5 to 6.5 (10% slurry in water)
Chemical Name or Material Aluminium oxide, weakly acidic, Brockmann I
Packaging Plastic bottle
Silicon Dioxide 0.03% max.
Sodium Oxide 0.4% max.
Molecular Formula Al2O3
Linear Formula Al2O3
Surface Area 135 to 165 m2/g
MDL Number MFCD00003424
CAS 1344-28-1
Fe2O3 0.03% max.

Specifications

Chemical Name or Material Aluminium oxide, weakly acidic, Brockmann I
CAS 1344-28-1
Purity Grade For chromatography
Molecular Formula Al2O3
Linear Formula Al2O3
Formula Weight 101.96
MDL Number MFCD00003424