missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminium oxide, weakly acidic, Brockmann I, For chromatography, ACROS Organics™

$117.46 - $117.46

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Linear Formula Al2O3
Formula Weight 101.96
MDL Number MFCD00003424
Chemical Name or Material Aluminium oxide, weakly acidic, Brockmann I
Grade For chromatography
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC392520010 SDS Thermo Fisher Scientific
AC392520010
Plastic bottle 1kg Each for $117.46
AC392520050 SDS Thermo Fisher Scientific
AC392520050
Plastic bottle 5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

for column chromatography
Description & Specifications

Chemical Identifiers

1344-28-1
Al2O3
MFCD00003424
For chromatography
Al2O3
101.96
Aluminium oxide, weakly acidic, Brockmann I

Specifications

101.96
1kg
5.5 to 6.5 (10% slurry in water)
Al2O3
0.03% max.
Al2O3
135 to 165 m2/g
1344-28-1
For chromatography
Powder
White to off-white
15, 350
MFCD00003424
Plastic bottle
0.03% max.
0.4% max.
Aluminium oxide, weakly acidic, Brockmann I