missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Aluminum oxide, for chromatography, neutral, Brockmann I, 40-300 μm, 60A, Thermo Scientific Chemicals
Aluminum oxide, Al2O3, CAS Number-1344-28-1, abrasit, abrarex, abramax, abramant, alundum, Aluminum oxide, alpha-alumina, fasertonerde, Aluminum oxide, aloxite, 2.5kg, 2980.0 deg.C, White, 3.97g/mL, 215-691-6, 300ppm max.
Supplier: Thermo Scientific Chemicals 366680010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for column chromatographySpecifications
2980.0°C | |
2050.0°C | |
1344-28-1 | |
6.5 to 7.5 (5% aq. suspension, stirred) | |
1 kg | |
Al2O3 | |
01,19; 02,17; 03,6; 04,8; 06,16; 07,5; 08,9; 09,8; 10,8; 11,22; 13,14; 15,9; 16,9 | |
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
dialuminum;oxygen(2-) | |
9989226 | |
Chromatography | |
3.97g/mL |
Aluminum oxide | |
Activated, Neutral, Brockmann I | |
White | |
Fine Powder | |
Al2O3 | |
MFCD00003424 | |
15, 350 | |
Solubility in water: insoluble | |
[O--].[O--].[O--].[Al+3].[Al+3] | |
101.96 | |
101.96 | |
Plastic bottle |
Chemical Identifiers
1344-28-1 | |
101.96 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
9989226 |
Al2O3 | |
MFCD00003424 | |
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
[O--].[O--].[O--].[Al+3].[Al+3] |
Safety and Handling
EINECSNumber : 215-691-6
RUO – Research Use Only