missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Aluminum oxide, for chromatography, basic, Brockmann I, 40-300μm, 60A, Thermo Scientific Chemicals
Aluminum oxide, Al2O3, CAS Number-1344-28-1, abramant, abramax, abrarex, abrasit, aloxite, alpha-alumina, Aluminum oxide, Aluminum oxide, alundum, fasertonerde, 1kg, Na2O <0.4%, 2977.0 deg.C, White, 3.9700g/mL, 215-691-6, 300ppm max.
$181.93 - $1522.04
Chemical Identifiers
CAS | 1344-28-1 |
---|---|
Molecular Formula | Al2O3 |
Molecular Weight (g/mol) | 101.96 |
MDL Number | MFCD00003424 |
InChI Key | PNEYBMLMFCGWSK-UHFFFAOYSA-N |
Synonym | aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum |
PubChem CID | 9989226 |
SMILES | [O--].[O--].[O--].[Al+3].[Al+3] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC189990010
|
Thermo Scientific Chemicals
189990010 |
1 kg | Plastic Bottle |
Each for $181.93
|
|
||||
AC189990051
|
Thermo Scientific Chemicals
189990051 |
5 kg | Plastic Bottle |
Each for $624.69
|
|
||||
AC189990250
|
Thermo Scientific Chemicals
189990250 |
25 kg | Fibre drum |
Each for $1,522.04
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatographyChemical Identifiers
1344-28-1 | |
101.96 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
9989226 |
Al2O3 | |
MFCD00003424 | |
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
[O--].[O--].[O--].[Al+3].[Al+3] |
Specifications
2977.0°C | |
1344-28-1 | |
9 to 10 (10% aq. suspension) | |
1 kg | |
Al2O3 | |
01,19; 02,17; 03,6; 04,8; 06,16; 07,5; 08,9; 09,8; 10,8; 11,22; 13,14; 15,9; 16,9 | |
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
dialuminium(3+) trioxidandiide | |
9989226 | |
Chromatography | |
3.97 | |
Aluminum oxide |
2030.0°C | |
White | |
Powder | |
Al2O3 | |
MFCD00003424 | |
15, 350 | |
Solubility in water: insoluble. Other solubilities: slightly sol. in alkaline sol.(forming hydroxides), practically insoluble in non-polar organic solvent, difficultly soluble in mineral acids | |
[O--].[O--].[O--].[Al+3].[Al+3] | |
101.96 | |
101.96 | |
Plastic Bottle | |
3.9700g/mL |
Safety and Handling
EINECSNumber : 215-691-6
RUO – Research Use Only