missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum oxide, for chromatography, acidic, Brockmann I, 40-300 μm, 60A, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 365750100

 View more versions of this product

Catalog No. AC365750100



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for column chromatography
Specifications

Specifications

2980.0°C
2030.0°C
1344-28-1
Powder
Al2O3
MFCD00003424
15,35
Solubility in water: insoluble. Other solubilities: slightly sol. in alkaline sol.(forming hydroxides), practically insoluble in non-polar organic solvent, difficultly soluble in mineral acids
[O--].[O--].[O--].[Al+3].[Al+3]
101.96
101.96
Plastic drum
Aluminum oxide
White
4.0 to 5.0 (5% aq. suspension, stirred)
10 kg
Al2O3
01,19; 02,17; 03,6; 04,8; 06,16; 07,5; 08,9; 09,8; 10,8; 11,22; 13,14; 15,9; 16,9
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminium(3+) trioxidandiide
9989226
Chromatography

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226
Al2O3
MFCD00003424
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
[O--].[O--].[O--].[Al+3].[Al+3]
Safety and Handling

Safety and Handling

EINECSNumber : 215-691-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only