missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminium nitrate nonahydrate, Acros Organics™

Manufacturer:  Acros Organics AC446340025

 View more versions of this product

Catalog No. AC446340025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Aluminium nitrate nonahydrate
AlH18N3O18
aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminium 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic
SWCIQHXIXUMHKA-UHFFFAOYSA-N
aluminum;trinitrate;nonahydrate
24567
98.0 to 102.0% (Complexometry)
0.005% max.
0.001% max.
0.002% max.
0.002% max.
Solubility in water: 64 g/100mL (25°C). Other solubilities: soluble in alcohol, slightly soluble in acetone, practically insoluble in ethyl acetate and, pyridine
73.0°C
7784-27-2
Al(NO3)3·9H2O
Solubility in water: 64 g/100mL (25°C). Other solubilities: soluble in alcohol, slightly soluble in acetone, practically insoluble in ethyl acetate and, pyridine
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
375.129
375.13
ACS Reagent
0.001% max.
0.005% max.
0.001% max.
0.005% max.
0.005% max.
2.5kg

Chemical Identifiers

7784-27-2
375.129
aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminium 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic
aluminum;trinitrate;nonahydrate
AlH18N3O18
SWCIQHXIXUMHKA-UHFFFAOYSA-N
24567
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
Safety and Handling

Safety and Handling

GHS H Statement Causes skin irritation. Causes serious eye irritation. May intensify fire; oxidizer.

GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

Warning

SDS Documents