Please login to your online account to display your discounted pricing

Aluminium isopropoxide, 98+%, ACROS Organics™


Quantity 5kg
Packaging Plastic bucket

Manufacturer: thermo fisher scientific  AC205170051

 View more versions of this product

Catalog No. AC205170051

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Formula Weight 204.25
Physical Form Crystalline Powder
Quantity 5kg
Color Pale yellow to yellow
Melting Point 129° to 136°C
Fieser 01,35; 03,10; 04,15; 05,14; 06,19; 08,15; 09,14; 11,29; 13,265; 15,16
Merck Index 15, 342
Linear Formula {(CH3)2CHO}3Al
Synonym AIP
Infrared Spectrum Authentic
CAS 555-31-7
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Flammable solid.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Aluminum (Al) 12.9 to 13.5 %
Packaging Plastic bucket
Chemical Name or Material Aluminium isopropoxide
Molecular Formula C9H21AlO3
MDL Number MFCD00008870
Assay Percent Range 98+%

Specifications

Chemical Name or Material Aluminium isopropoxide
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C9H21AlO3
Linear Formula {(CH3)2CHO}3Al
Formula Weight 204.25
MDL Number MFCD00008870
Synonym AIP
Accessories

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable solid.

SDS & Safety Documents