Learn More
Aluminum chloride hexahydrate, 99%, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 217475000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidSpecifications
Aluminum chloride hexahydrate | |
500 g | |
AlCl3H12O6 | |
MFCD00149134 | |
15,333 | |
JGDITNMASUZKPW-UHFFFAOYSA-K | |
aluminium(3+) hexahydrate trichloride | |
241.43 | |
Pure | |
2.39 |
7784-13-6 | |
98.5% min. (Argentometry) | |
AlCl3·6H2O | |
01,24; 02,21; 03,7; 04,10; 05,10; 06,17; 08,13; 09,11; 17,15 | |
Solubility in water: 477g/L (20°C). Other solubilities: soluble in alcohol, ether, glycerol and propylene | |
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-] | |
241.42 | |
99% | |
Glass bottle | |
2.3900g/mL |
Chemical Identifiers
7784-13-6 | |
241.42 | |
JGDITNMASUZKPW-UHFFFAOYSA-K |
AlCl3H12O6 | |
MFCD00149134 | |
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-] |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
If skin irritation occurs: Get medical advice/attention.
Wash face,hands and any ex
GHS Signal Word: Warning
RUO – Research Use Only