missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Aluminium chloride hexahydrate, 99%, pure, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 217470100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Lewis acidSpecifications
Aluminium chloride hexahydrate | |
10kg | |
AlCl3H12O6 | |
MFCD00149134 | |
15,333 | |
JGDITNMASUZKPW-UHFFFAOYSA-K | |
aluminium(3+) hexahydrate trichloride | |
241.43 | |
Pure | |
2.39 |
7784-13-6 | |
98.5% min. (Argentometry) | |
AlCl3·6H2O | |
01,24; 02,21; 03,7; 04,10; 05,10; 06,17; 08,13; 09,11; 17,15 | |
Solubility in water: 477g/L (20°C). Other solubilities: soluble in alcohol,ether,glycerol and propylene | |
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-] | |
241.42 | |
99% | |
Plastic drum | |
2.3900g/mL |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
If skin irritation occurs: Get medical advice/attention.
Wash face,hands and any ex
GHS Signal Word: Warning
RUO â Research Use Only