Learn More
Aluminum chloride, 99.999%, (trace metal basis), for analysis, anhydrous, powder, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 364810100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis, Lewis acidSpecifications
Aluminum chloride | |
7446-70-0 | |
Powder | |
99.999% | |
AlCl3 | |
01,24; 02,21; 03,7; 04,10; 05,10; 06,17; 07,7; 08,13; 09,11; 10,9; 11,25; 12,26; 13,15; 14,20; 15,10; 16,10; 17,15 | |
aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum | |
VSCWAEJMTAWNJL-UHFFFAOYSA-K | |
aluminum(3+) trichloride | |
133.34 | |
Trace Metal Basis | |
2.44 |
194.0°C | |
White | |
10 g | |
AlCl3 | |
MFCD00003422 | |
15,333 | |
Solubility in water: reacts. Other solubilities: freely miscible with benzene, nitrobenzene,, benzophenone, carbon tetrachloride and chloroform | |
[Al+3].[Cl-].[Cl-].[Cl-] | |
133.33 | |
99.999% | |
Glass Bottle | |
2.4400g/mL |
Chemical Identifiers
7446-70-0 | |
133.33 | |
VSCWAEJMTAWNJL-UHFFFAOYSA-K | |
aluminum(3+) trichloride |
AlCl3 | |
MFCD00003422 | |
aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum | |
[Al+3].[Cl-].[Cl-].[Cl-] |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
EINECSNumber : 231-208-1
RUO – Research Use Only